accessibility menu, dialog, popup

UserName

CH3SOOClBr

CAS RN 72256-93-0

IUPAC Name: 4-bromo-3-methylbenzene-1-sulfonyl chloride
Synonyms: 4-bromo-3-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: JRNDXUKMWFVEIC-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)