accessibility menu, dialog, popup

UserName

CAS RN 7159-36-6

NOHO

CAS RN 7159-36-6

IUPAC Name: isoquinoline-4-carboxylic acid
Synonyms: isoquinoline-4-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: MCVMLYSLPCECGO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results