accessibility menu, dialog, popup

UserName

CAS RN 71550-12-4

NH2H2C

CAS RN 71550-12-4

IUPAC Name: prop-2-en-1-amine
Synonyms: allylamine
Molecular Weight (g/mol): 57.1
Molecular Formula: C3H7N
InChi Key: VVJKKWFAADXIJK-UHFFFAOYSA-N
SMILES: NCC=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)