CAS RN 71208-06-5
CAS RN 71208-06-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lewis X Trisaccharide, TRC
CAS: 71208-06-5 Molecular Formula: C20 H35 N O15 Molecular Weight (g/mol): 529.49 IUPAC Name: N-[(3R,4R,5S,6R)-2-hydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl]acetamide SMILES: C[C@@H]1O[C@@H](O[C@@H]2[C@@H](NC(=O)C)C(O)O[C@H](CO)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O