accessibility menu, dialog, popup

UserName

CAS RN 71189-15-6

ONCO

CAS RN 71189-15-6

IUPAC Name: 2-(isocyanatomethyl)furan
Synonyms: 2-(isocyanatomethyl)furan
Molecular Weight (g/mol): 123.11
Molecular Formula: C6H5NO2
InChi Key: UIADMYLYGJYUSQ-UHFFFAOYSA-N
SMILES: O=C=NCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results