CAS RN 7093-67-6
CAS RN 7093-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pentaglycine, TRC
CAS: 7093-67-6 Molecular Formula: C10H17N5O6 Molecular Weight (g/mol): 303.27 IUPAC Name: 2-[[2-[[2-[[2-[(2-aminoacetyl)amino]acetyl]amino]acetyl]amino]acetyl]amino]acetic acid SMILES: NCC(=O)NCC(=O)NCC(=O)NCC(=O)NCC(=O)O