accessibility menu, dialog, popup

UserName

CAS RN 7059-70-3

CAS RN 7059-70-3

IUPAC Name: 2-[3,5-bis(naphthalen-2-yl)phenyl]naphthalene
Synonyms: 2-[3,5-bis(naphthalen-2-yl)phenyl]naphthalene
Molecular Weight (g/mol): 456.59
Molecular Formula: C36H24
InChi Key: AWIOMAWDKDFGDP-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C(C=C2C=C1)C1=CC(=CC(=C1)C1=CC2=CC=CC=C2C=C1)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)