accessibility menu, dialog, popup

UserName

CAS RN 705-79-3

OHOFCl

CAS RN 705-79-3

IUPAC Name: 2-(3-chloro-4-fluorophenyl)acetic acid
Synonyms: (3-chloro-4-fluorophenyl)acetic acid
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: WGODGMOMOMNTRN-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results