accessibility menu, dialog, popup

UserName

CAS RN 70060-13-8

OOSF

CAS RN 70060-13-8

IUPAC Name: 5-fluoro-1-benzothiophene-2-carboxylate
Synonyms: 5-fluoro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 195.19
Molecular Formula: C9H4FO2S
InChi Key: PLVPMOSTGNZKQQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(F)=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results