CAS RN 698-19-1
IUPAC Name:
[(2-bromophenyl)methyl](methyl)azanium
Synonyms:
[(2-bromophenyl)methyl](methyl)azanium
Molecular Weight (g/mol):
201.09
Molecular Formula:
C8H11BrN
InChi Key:
TUADRPBKJHMHDH-UHFFFAOYSA-O
SMILES:
C[NH2+]CC1=CC=CC=C1Br
1
–
1
of
1
results
1
2-Bromo-N-methylbenzylamine, 95%, Thermo Scientific™
CAS: 698-19-1 Molecular Formula: C8H10BrN Molecular Weight (g/mol): 200.079 MDL Number: MFCD05262953 InChI Key: TUADRPBKJHMHDH-UHFFFAOYSA-N Synonym: 2-bromobenzyl methylamine,n-methyl-2-bromobenzylamine,2-bromo-n-methylbenzylamine,1-2-bromophenyl-n-methylmethanamine,2-bromophenyl methyl methyl amine,benzenemethanamine, 2-bromo-n-methyl,n-methyl-o-bromobenzylamine,acmc-20ao1g,2-bromo-n-methyl benzylamine,2-bromo-benzyl-methyl-amine PubChem CID: 522360 IUPAC Name: 1-(2-bromophenyl)-N-methylmethanamine SMILES: CNCC1=CC=CC=C1Br