accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 6975-60-6

IUPAC Name: 1-(furan-2-yl)propan-2-one
Synonyms: 2-propanone, 1-(2-furanyl)-
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: IQOJTGSBENZIOL-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results