accessibility menu, dialog, popup

UserName

CAS RN 69739-34-0

H3CCH3H3CSiCH3H3COSOOFFF

CAS RN 69739-34-0

IUPAC Name: tert-butyldimethylsilyl trifluoromethanesulfonate
Synonyms: tert-butyldimethylsilyl trifluoromethanesulfonate
Molecular Weight (g/mol): 264.33
Molecular Formula: C7H15F3O3SSi
InChi Key: WLLIXJBWWFGEHT-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)