accessibility menu, dialog, popup

UserName

CAS RN 6971-44-4

IUPAC Name: methyl[(pyridin-4-yl)methyl]amine
Synonyms: methyl(pyridin-4-ylmethyl)amine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: DNBWGFKLIBQQSL-UHFFFAOYSA-N
SMILES: CNCC1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)