accessibility menu, dialog, popup

UserName

CAS RN 697-90-5

ClH2NICl

CAS RN 697-90-5

IUPAC Name: 2,4-dichloro-6-iodoaniline
Synonyms: 2,4-dichloro-6-iodoaniline
Molecular Weight (g/mol): 287.91
Molecular Formula: C6H4Cl2IN
InChi Key: YJRSMJTVXWBFJJ-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results