accessibility menu, dialog, popup

UserName

NFN

CAS RN 696-42-4

IUPAC Name: 5-fluoropyridine-3-carbonitrile
Synonyms: 5-fluoropyridine-3-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: LQJWCPZRNHAVJD-UHFFFAOYSA-N
SMILES: FC1=CN=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results