accessibility menu, dialog, popup

UserName

CAS RN 69411-06-9

FH2NFCl

CAS RN 69411-06-9

IUPAC Name: 4-chloro-2,6-difluoroaniline
Synonyms: 4-chloro-2,6-difluoroaniline
Molecular Weight (g/mol): 163.55
Molecular Formula: C6H4ClF2N
InChi Key: AKDPHEYBBWDWCM-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results