CAS RN 69388-79-0
CAS RN 69388-79-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sulbactam Pivoxil, TRC
CAS: 69388-79-0 Molecular Formula: C14 H21 N O7 S Molecular Weight (g/mol): 347.38 Synonym: (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (2,2-Dimethyl-1-oxopropoxy)methyl Ester 4,4-Dioxide,(Pivaloyloxy)methyl Penicillanate S,S-Dioxide,CP 47904,(2S-cis)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (2,2-dimethyl-1-oxopropoxy)methyl Ester 4,4-Dioxide IUPAC Name: 2,2-dimethylpropanoyloxymethyl (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate SMILES: CC(C)(C)C(=O)OCOC(=O)[C@@H]1N2[C@@H](CC2=O)S(=O)(=O)C1(C)C