accessibility menu, dialog, popup

UserName

CAS RN 69113-32-2

NOHH2NF(E/Z)

CAS RN 69113-32-2

IUPAC Name: 4-fluoro-N'-hydroxybenzene-1-carboximidamide
Synonyms: 4-fluoro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: OSUPWUQRPLIJKX-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=C(F)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results