accessibility menu, dialog, popup

UserName

CAS RN 69097-20-7

H3CSiCH3H3COOSiCH3H3CCH3OSiCH3H3CCH3

CAS RN 69097-20-7

IUPAC Name: 2,2,7,7-tetramethyl-4-[(trimethylsilyl)oxy]-3,6-dioxa-2,7-disilaoct-4-ene
Synonyms: 2,2,7,7-tetramethyl-4-[(trimethylsilyl)oxy]-3,6-dioxa-2,7-disilaoct-4-ene
Molecular Weight (g/mol): 292.6
Molecular Formula: C11H28O3Si3
InChi Key: FCZGHPGTZRTDNN-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results