CAS RN 690645-98-8
IUPAC Name:
4-[(3-chlorobenzenesulfonamido)methyl]benzoic acid
Synonyms:
4-[(3-chlorobenzenesulfonamido)methyl]benzoic acid
Molecular Weight (g/mol):
325.76
Molecular Formula:
C14H12ClNO4S
InChi Key:
CTNSYQWHQNQSIT-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(CNS(=O)(=O)C2=CC(Cl)=CC=C2)C=C1
1
–
1
of
1
results
1
4-(3-Chlorophenylsulfonylaminomethyl)benzoic acid, 96%, Thermo Scientific™
CAS: 690645-98-8 Molecular Formula: C14H12ClNO4S Molecular Weight (g/mol): 325.763 MDL Number: MFCD04154033 InChI Key: CTNSYQWHQNQSIT-UHFFFAOYSA-N Synonym: 4-3-chlorophenylsulfonylaminomethyl benzoic acid,4-3-chlorophenyl sulfonamido methyl benzoic acid,4-3-chlorobenzenesulfonamido methyl benzoic acid,4-3-chlorophenylsulfonamido methyl benzoic acid,4-3-chlorophenyl sulfonylamino methyl benzoic acid,4-3-chlorophenyl sulfonyl amino methyl benzoic acid PubChem CID: 852336 IUPAC Name: 4-[[(3-chlorophenyl)sulfonylamino]methyl]benzoic acid SMILES: C1=CC(=CC(=C1)Cl)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)O