accessibility menu, dialog, popup

UserName

CAS RN 68867-20-9

H3CNSI

CAS RN 68867-20-9

IUPAC Name: 6-iodo-2-methyl-1,3-benzothiazole
Synonyms: 6-iodo-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 275.11
Molecular Formula: C8H6INS
InChi Key: YVXGKAZJAUWMPM-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(I)C=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results