accessibility menu, dialog, popup

UserName

HOOOO

CAS RN 68858-21-9

IUPAC Name: 2-[4-(hydroxymethyl)phenoxy]acetate
Synonyms: 2-[4-(hydroxymethyl)phenoxy]acetate
Molecular Weight (g/mol): 181.17
Molecular Formula: C9H9O4
InChi Key: VUCNQOPCYRJCGQ-UHFFFAOYSA-M
SMILES: OCC1=CC=C(OCC([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)