accessibility menu, dialog, popup

UserName

H3C(S)OHHNOCH3OO

CAS RN 68858-20-8

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular Weight (g/mol): 339.39
Molecular Formula: C20H21NO4
InChi Key: UGNIYGNGCNXHTR-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)