accessibility menu, dialog, popup

UserName

CH3NBr

CAS RN 68470-59-7

IUPAC Name: 2-(bromomethyl)-6-methylpyridine
Synonyms: 2-(bromomethyl)-6-methylpyridine
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: WJFDCFHWFHCLIW-UHFFFAOYSA-N
SMILES: CC1=NC(CBr)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results