accessibility menu, dialog, popup

UserName

(S)(R)(R)O(R)(S)(R)(R)O(R)NONH2HOOHOPOOOOHONNNNNH2OPOOKPOHOHO

CAS RN 68141-45-7

IUPAC Name: potassium 1-[5-({[({[5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Synonyms: potassium 1-(5-{[({[5-(6-aminopurin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methyl phosphonato}oxy(oxido)phosphoryl)oxy]methyl}-3,4-dihydroxyoxolan-2-yl)-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular Weight (g/mol): 781.5
Molecular Formula: C21H27KN7O17P3
InChi Key: YUTBYWSDZWYADH-UHFFFAOYNA-M
SMILES: [K+].NC(=O)C1=C[N+](=CC=C1)C1OC(COP([O-])(=O)OP([O-])(=O)OCC2OC(C(OP(O)(O)=O)C2O)N2C=NC3=C(N)N=CN=C23)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results