CAS RN 681-99-2
IUPAC Name:
tributyl(isocyanato)stannane
Synonyms:
tributyl(isocyanato)stannane
Molecular Weight (g/mol):
334.09
Molecular Formula:
C13H29NOSn
InChi Key:
KYQISWRYYMJYEP-UHFFFAOYSA-N
SMILES:
N=C=O.CCCC[SnH](CCCC)CCCC
1
–
1
of
1
results
1
Tri-n-butyltin isocyanate, 97%, Thermo Scientific™
CAS: 681-99-2 Molecular Formula: C13H27NOSn Molecular Weight (g/mol): 332.075 MDL Number: MFCD08272321 InChI Key: YGUNDMHXMXDGII-UHFFFAOYSA-N Synonym: tributyltin isocyanate,tributyl isocyanato stannane,stannane, isocyanato tributyl,tri-n-butyltin isocyanate,tributylstannyl isocyanate,tributyltin isothiocyanate,stannane, tributylisocyanato,tin, tributyl-, isothiocyanate,tri-n-butylisocyanatotin,acmc-20ajf1 PubChem CID: 120323 IUPAC Name: tributyl(isocyanato)stannane SMILES: CCCC[Sn](CCCC)(CCCC)N=C=O