accessibility menu, dialog, popup

UserName

BrONN

CAS RN 68047-37-0

IUPAC Name: 2-(4-bromophenyl)-5-(naphthalen-1-yl)-1,3,4-oxadiazole
Synonyms: 2-(4-bromophenyl)-5-(naphthalen-1-yl)-1,3,4-oxadiazole
Molecular Weight (g/mol): 351.2
Molecular Formula: C18H11BrN2O
InChi Key: VZEIYIPOCFGTPV-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NN=C(O1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results