accessibility menu, dialog, popup

UserName

CAS RN 6783-05-7

HOBOH(E)

CAS RN 6783-05-7

IUPAC Name: [(1E)-2-phenylethenyl]boronic acid
Synonyms: (1E)-2-phenylethenylboronic acid
Molecular Weight (g/mol): 147.97
Molecular Formula: C8H9BO2
InChi Key: VKIJXFIYBAYHOE-VOTSOKGWSA-N
SMILES: OB(O)\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results