CAS RN 67748-61-2
IUPAC Name:
1,3-benzothiazole-2-carbonyl chloride
Synonyms:
1,3-benzothiazole-2-carbonyl chloride
Molecular Weight (g/mol):
197.64
Molecular Formula:
C8H4ClNOS
InChi Key:
AOIGQLLPWDXVGB-UHFFFAOYSA-N
SMILES:
ClC(=O)C1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
Benzothiazole-2-carbonyl chloride, 95%
CAS: 67748-61-2 Molecular Formula: C8H4ClNOS Molecular Weight (g/mol): 197.636 MDL Number: MFCD03659697 InChI Key: AOIGQLLPWDXVGB-UHFFFAOYSA-N Synonym: benzothiazole-2-carbonyl chloride,2-benzothiazolecarbonyl chloride,benzo d thiazole-2-carbonyl chloride,acmc-1b36m,2-benzothiazolecarbonylchloride,benzothiazol-2-carbonyl chloride,benzothiazole-2-carbonyl chloride .,2-benzothiazolecarbonyl chloride 9ci,benzothiazole-2-carboxylic acid chloride PubChem CID: 2776254 IUPAC Name: 1,3-benzothiazole-2-carbonyl chloride SMILES: C1=CC=C2C(=C1)N=C(S2)C(=O)Cl