CAS RN 675126-09-7
CAS RN 675126-09-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,4R)-N-Desmethyl Sertraline Hydrochloride, TRC
CAS: 675126-09-7 Molecular Formula: C16 H15 Cl2 N . Cl H Molecular Weight (g/mol): 328.66 Synonym: 1-Naphthalenamine, 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-, hydrochloride (1:1), (1R,4R)-,1-Naphthalenamine, 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-, hydrochloride, (1R,4R)- (9CI),(1R,4R)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride,(1R,4R)-N-Desmethylsertraline hydrochloride,(1R,4R)-N-Desmethyl sertraline hydrochloride,(1R,4R)-Norsertraline hydrochloride IUPAC Name: (1R,4R)-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride SMILES: Cl.N[C@@H]1CC[C@H](c2ccc(Cl)c(Cl)c2)c3ccccc13