CAS RN 6746-81-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycidyl 4-Toluenesulfonate, TRC
CAS: 6746-81-2 Molecular Formula: C10H12O4S Molecular Weight (g/mol): 228.26 Synonym: (±)-Glycidyl Tosylate,2,3-Epoxy-1-propyl p-Toluenesulfonate;2,3-Epoxypropyl Tosylate,Glycidol p-Toluenesulfonate,Glycidyl p-Toluenesulfonate,Glycidyl Tosylate,NSC 143322,Toluene-4-sulfonic Acid Oxiranylmethyl Ester,4-Methylbenzenesulfonate Oxiranemethanol; IUPAC Name: oxiran-2-ylmethyl 4-methylbenzenesulfonate SMILES: Cc1ccc(cc1)S(=O)(=O)OCC2CO2