accessibility menu, dialog, popup

UserName

H3COON

CAS RN 67284-56-4

IUPAC Name: 4'-cyano-[1,1'-biphenyl]-4-yl 4-ethylcyclohexane-1-carboxylate
Synonyms: 4'-cyano-[1,1'-biphenyl]-4-yl 4-ethylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 333.43
Molecular Formula: C22H23NO2
InChi Key: OYZITXKNUGHHKV-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C(=O)OC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results