CAS RN 67-96-9
CAS RN 67-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydro Tachysterol, TRC
CAS: 67-96-9 Molecular Formula: C28 H46 O Molecular Weight (g/mol): 398.66 Synonym: Dihydrotachysterol,(5E,7E,22E)-9,10-Seco-10alpha-ergosta-5,7,22-trien-3beta-ol IUPAC Name: (1S,3E,4S)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylcyclohexan-1-ol SMILES: CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@@H](O)CC[C@@H]3C)\CCC[C@]12C