accessibility menu, dialog, popup

UserName

NBrOHOF

CAS RN 669066-91-5

IUPAC Name: 5-bromo-3-fluoropyridine-2-carboxylic acid
Synonyms: 5-bromo-3-fluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: JXHUXOAJAYWMHK-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(Br)C=C1F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)