accessibility menu, dialog, popup

UserName

CAS RN 667-27-6

H3COOFFBr

CAS RN 667-27-6

IUPAC Name: ethyl 2-bromo-2,2-difluoroacetate
Synonyms: ethyl 2-bromo-2,2-difluoroacetate
Molecular Weight (g/mol): 202.98
Molecular Formula: C4H5BrF2O2
InChi Key: IRSJDVYTJUCXRV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)Br
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)