accessibility menu, dialog, popup

UserName

CAS RN 66659-20-9

NSNH2OHOHCl

CAS RN 66659-20-9

IUPAC Name: 2-(2-amino-1,3-thiazol-4-yl)acetic acid hydrochloride
Synonyms: (2-amino-1,3-thiazol-4-yl)acetic acid hydrochloride
Molecular Weight (g/mol): 194.63
Molecular Formula: C5H7ClN2O2S
InChi Key: ZEWJJZKVQOMYKJ-UHFFFAOYSA-N
SMILES: Cl.NC1=NC(CC(O)=O)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results