accessibility menu, dialog, popup

UserName

ClO

CAS RN 6640-25-1

IUPAC Name: (4-chlorophenyl)(cyclopropyl)methanone
Synonyms: (4-chlorophenyl)(cyclopropyl)methanone
Molecular Weight (g/mol): 180.63
Molecular Formula: C10H9ClO
InChi Key: OPSFCTBBDIDFJM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results