accessibility menu, dialog, popup

UserName

CAS RN 6625-96-3

ONOHNO

CAS RN 6625-96-3

IUPAC Name: 5-nitro-1H-indole-3-carbaldehyde
Synonyms: 5-nitro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: PHKYMSLVWLYDKP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC=C(C=O)C2=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)