CAS RN 662138-96-7
IUPAC Name:
6-phenyl-2-(pyridin-2-yl)-1,3,6,2-dioxazaborocane; propan-2-ol
Synonyms:
6-phenyl-2-(pyridin-2-yl)-1,3,6,2-dioxazaborocane; isopropyl alcohol
Molecular Weight (g/mol):
328.22
Molecular Formula:
C18H25BN2O3
InChi Key:
XMHAHEGWYGUDFM-UHFFFAOYSA-N
SMILES:
CC(C)O.C1CN(CCOB(O1)C1=CC=CC=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact composition
CAS: 662138-96-7 Molecular Formula: C15H17BN2O2 Molecular Weight (g/mol): 268.12 MDL Number: MFCD06798116 InChI Key: FZYLTWAWSRCZPG-UHFFFAOYSA-N Synonym: 2-pyridineboronic acid 2,2'-phenylimino diethanol ester,pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol,pubchem6434,6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol PubChem CID: 49761666 SMILES: C1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1