accessibility menu, dialog, popup

UserName

CAS RN 6609-54-7

H3CSN

CAS RN 6609-54-7

IUPAC Name: 2-(methylsulfanyl)benzonitrile
Synonyms: 2-(methylsulfanyl)benzonitrile
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: PXZSANDJGNKIIA-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)