accessibility menu, dialog, popup

UserName

CAS RN 660848-57-7

H3COOFFFNH2

CAS RN 660848-57-7

IUPAC Name: 2-methoxy-5-(trifluoromethoxy)aniline
Synonyms: 2-methoxy-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 207.15
Molecular Formula: C8H8F3NO2
InChi Key: MKAPTOAKEPIGOA-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results