accessibility menu, dialog, popup

UserName

CAS RN 655-15-2

FOBr

CAS RN 655-15-2

IUPAC Name: 2-bromo-1-(2-fluorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(2-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: QDNWNJSLWKHNTM-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)CBr
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)