accessibility menu, dialog, popup

UserName

NH3O

CAS RN 65412-03-5

IUPAC Name: 2-(oxan-4-yl)ethan-1-aminium
Synonyms: 2-(oxan-4-yl)ethanaminium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: BZMADPOGYCRPAI-UHFFFAOYSA-O
SMILES: [NH3+]CCC1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)