accessibility menu, dialog, popup

UserName

CAS RN 654055-01-3

IUPAC Name: 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one hydrate
Synonyms: morin (flavonol)
Molecular Weight (g/mol): 320.25
Molecular Formula: C15H12O8
InChi Key: MYUBTSPIIFYCIU-UHFFFAOYSA-N
SMILES: O.OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results