accessibility menu, dialog, popup

UserName

ClNBr

CAS RN 65340-70-7

IUPAC Name: 6-bromo-4-chloroquinoline
Synonyms: 6-bromo-4-chloroquinoline
Molecular Weight (g/mol): 242.5
Molecular Formula: C9H5BrClN
InChi Key: KJILYZMXTLCPDQ-UHFFFAOYSA-N
SMILES: ClC1=C2C=C(Br)C=CC2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results