accessibility menu, dialog, popup

UserName

CAS RN 653-34-9

FFFCH2FF

CAS RN 653-34-9

IUPAC Name: 1-ethenyl-2,3,4,5,6-pentafluorobenzene
Synonyms: 1-ethenyl-2,3,4,5,6-pentafluorobenzene
Molecular Weight (g/mol): 194.1
Molecular Formula: C8H3F5
InChi Key: LVJZCPNIJXVIAT-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C=C)C(F)=C1F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)