accessibility menu, dialog, popup

UserName

CAS RN 651358-83-7

CH3H3COBOH3CH3CBrN

CAS RN 651358-83-7

IUPAC Name: 2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 283.96
Molecular Formula: C11H15BBrNO2
InChi Key: OAVRLCKBKDMGGW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results