accessibility menu, dialog, popup

UserName

CH3NNH3CCl

CAS RN 65039-09-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium
Synonyms: 1-ethyl-3-methylimidazolium
Molecular Weight (g/mol): 111.17
Molecular Formula: C6H11N2
InChi Key: NJMWOUFKYKNWDW-UHFFFAOYSA-N
SMILES: CCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)