accessibility menu, dialog, popup

UserName

H3COOCH3CH3NCO

CAS RN 64505-10-8

IUPAC Name: ethyl 2-isocyanato-4-methylpentanoate
Synonyms: ethyl 2-isocyanato-4-methylpentanoate
Molecular Weight (g/mol): 185.22
Molecular Formula: C9H15NO3
InChi Key: SSTQICMSRVMLQN-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CC(C)C)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)