accessibility menu, dialog, popup

UserName

CAS RN 64201-64-5

ON

CAS RN 64201-64-5

IUPAC Name: hydrate quinolin-1-ium-1-olate
Synonyms: quinoline-1-oxide hydrate
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: CUSWDTBBCIXCRB-UHFFFAOYSA-N
SMILES: O.[O-][N+]1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results